Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031237.1 | NP_112516.1 | 4 | 411 | ||
XM_006233333.1 | XP_006233395.1 | 5 | 403 | |||
XM_006233334.1 | XP_006233396.1 | 5 | 550 | |||
XM_006233335.1 | XP_006233397.1 | 5 | 357 | |||
XM_006233336.2 | XP_006233398.1 | 5 | 403 | |||
XM_006233337.1 | XP_006233399.1 | 5 | 550 | |||
XM_006233338.1 | XP_006233400.1 | 5 | 388 | |||
GenBank mRNA | AB006852.1 | - | 4 | 411 | ||
AY724500.1 | - | 5 | 358 | |||
BC072696.1 | - | 5 | 334 | 7105617 | ||
FQ226278.1 | - | 5 | 387 | |||
FQ233355.1 | - | 5 | 378 | |||
U13175.1 | - | 4 | 265 |