Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3gl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031238.2 | NP_112517.1 | 9 | 1284 | ||
XM_006229480.2 | XP_006229542.1 | 9 | 1072 | |||
XM_006229481.2 | XP_006229543.1 | 9 | 1165 | |||
XM_006229483.2 | XP_006229545.1 | 9 | 1271 | |||
XM_006229485.1 | XP_006229547.1 | 7 | 815 | |||
XM_006229486.2 | XP_006229548.1 | 7 | 739 | |||
GenBank mRNA | AB008159.2 | - | 6 | 729 | ||
AF009604.1 | - | 7 | 768 | |||
AF227439.1 | - | 9 | 1287 | |||
BC078800.1 | - | 9 | 1287 | 7135194 |