Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191110.1 | NP_001178039.1 | 1 | 596 | ||
XM_006233192.2 | XP_006233254.1 | 1 | 1126 |
Set Membership: |
G-protein coupled receptor |