Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031322.3 | NP_112612.2 | 27 | 3858 | ||
XM_008762038.1 | XP_008760260.1 | 27 | 4070 | |||
GenBank mRNA | AB011533.1 | - | 14 | 2024 | ||
AB073317.3 | - | 27 | 3858 |