Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031356.1 | NP_112646.1 | 12, 13 | 1387 | ||
GenBank mRNA | AB041723.1 | - | 12, 13 | 1403 | ||
AF262320.1 | - | 12, 13 | 1309 | |||
AF375656.1 | - | 12, 13 | 1313 | |||
AY787821.1 | - | 12, 13 | 1328 | |||
BC072697.1 | - | 12, 13 | 1345 | 7105761 |