Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agxt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031835.2 | NP_114023.1 | 9, 10 | 995 | ||
XM_006232069.2 | XP_006232131.1 | 9, 10 | 1063 | |||
GenBank mRNA | AB002584.1 | - | 9, 10 | 966 | ||
BC081765.1 | - | 9, 10 | 995 | 7106995 | ||
D38100.1 | - | 9, 10 | 1010 | |||
FQ219625.1 | - | 1, 2 | 76 |