Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsfl1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031981.2 | NP_114187.1 | 8 | 1032 | ||
XM_006235254.2 | XP_006235316.1 | 9 | 1005 | |||
GenBank mRNA | AB002086.1 | - | 8 | 1032 | ||
BC072464.1 | - | 8 | 929 | 7096463 | ||
FQ212982.1 | - | 1 | 16 | |||
FQ228953.1 | - | 9 | 970 | |||
Y10769.1 | - | 8 | 929 |