Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031982.1 | NP_114188.1 | 13 | 1960 | ||
XM_008767882.1 | XP_008766104.1 | 12 | 1967 | |||
GenBank mRNA | AB015231.3 | - | 1211 | |||
AB040873.1 | - | 12 | 1880 | |||
AB041029.1 | - | 11 | 1700 | |||
AF029310.1 | - | 13 | 1960 | |||
AF158248.2 | - | 7 | 1293 | |||
LC008303.1 | - | 8 | 1200 |