Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdcbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031986.1 | NP_114192.1 | 8 | 847 | ||
GenBank mRNA | AF248548.1 | - | 8 | 847 | ||
AJ292243.1 | - | 7 | 790 | |||
BC064651.1 | - | 8 | 815 | 5598152 |