Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kalrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032062.2 | NP_114451.2 | 53 | 7650 | ||
GenBank mRNA | AF232669.1 | - | 52 | 7620 |