Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032073.1 | NP_114462.1 | 6 | 942 | ||
GenBank mRNA | AJ133685.1 | - | 6 | 942 | ||
AY043394.1 | - | 5 | 682 | |||
U92655.1 | - | 5 | 454 |