Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032077.1 | NP_114466.1 | 2, 3 | 157 | ||
GenBank mRNA | BC091403.1 | - | 2, 3 | 198 | 7324332 | |
CO570909.1 | - | 2, 3 | 157 | |||
FQ209439.1 | - | 2, 3 | 175 | |||
FQ211027.1 | - | 2, 3 | 168 | |||
FQ211041.1 | - | 2, 3 | 211 | |||
FQ218888.1 | - | 2, 3 | 168 | |||
FQ219478.1 | - | 2, 3 | 160 | |||
FQ219506.1 | - | 2, 3 | 168 | |||
FQ219654.1 | - | 2, 3 | 160 | |||
FQ219776.1 | - | 2, 3 | 168 | |||
FQ223824.1 | - | 1, 2 | 68 | |||
U94856.1 | - | 2, 3 | 115 |