Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032077.1 | NP_114466.1 | 7 | 742 | ||
GenBank mRNA | BC091403.1 | - | 7 | 783 | 7324332 | |
CO562983.1 | - | 1 | 36 | |||
FQ209439.1 | - | 7 | 760 | |||
FQ209632.1 | - | 4 | 438 | |||
FQ211027.1 | - | 7 | 753 | |||
FQ211041.1 | - | 7 | 796 | |||
FQ218360.1 | - | 7 | 758 | |||
FQ218888.1 | - | 7 | 753 | |||
FQ219506.1 | - | 7 | 753 | |||
FQ219654.1 | - | 7 | 745 | |||
FQ219750.1 | - | 7 | 747 | |||
FQ219776.1 | - | 7 | 754 | |||
FQ223824.1 | - | 6 | 653 | |||
U94856.1 | - | 7 | 700 |