Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053354.3 | NP_445806.3 | 19 | 1672 | ||
GenBank mRNA | AB012214.1 | - | 19 | 1656 | ||
CB712236.1 | - | 23 | ||||
EV775426.1 | - | 303 |