Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053360.2 | NP_445812.1 | 13, 14 | 1657 | ||
XM_006256924.2 | XP_006256986.2 | 13, 14 | 1599 | |||
XM_006256929.2 | XP_006256991.2 | 11, 12 | 1425 | |||
XM_006256930.2 | XP_006256992.1 | 8, 9 | 1018 | |||
XM_006256931.1 | XP_006256993.1 | 15, 16 | 1950 | |||
XM_008773199.1 | XP_008771421.1 | 2, 3 | 208 | |||
XM_008773200.1 | XP_008771422.1 | 2, 3 | 148 | |||
XM_008773201.1 | XP_008771423.1 | 2, 3 | 104 | |||
XM_008773202.1 | XP_008771424.1 | 2, 3 | 129 | |||
GenBank mRNA | AF255885.1 | - | 1, 2 | 375 | ||
AF255886.1 | - | 9, 10 | 894 | |||
AF255887.1 | - | 8, 9 | 788 | |||
AF255888.1 | - | 13, 14 | 1575 | |||
BC070877.1 | - | 13, 14 | 1657 | 7096160 | ||
FQ226336.1 | - | 5, 6 | 473 |