Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053364.1 | NP_445816.1 | 5, 6 | 608 | ||
GenBank mRNA | AF139809.1 | - | 5, 6 | 608 | ||
AF226609.1 | - | 5, 6 | 580 | |||
FQ219949.1 | - | 5, 6 | 604 | |||
FQ220120.1 | - | 5, 6 | 555 |