Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257113.2 | XP_006257175.1 | 24 | 2948 | ||
XM_006257114.1 | XP_006257176.1 | 24 | 2964 | |||
XM_343797.7 | XP_343798.2 | 23 | 3063 | |||
GenBank mRNA | AF155826.1 | - | 2 | 185 |