Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053416.2 | NP_445868.2 | 4 | 677 | ||
XM_006234108.2 | XP_006234170.2 | 5 | 1015 | |||
XM_008761777.1 | XP_008759999.1 | 5 | 715 | |||
XM_008761778.1 | XP_008760000.1 | 5 | 715 | |||
GenBank mRNA | CA512540.1 | - | 1 | 106 |