Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749609.2 | XP_003749657.1 | 6, 7 | 676 | ||
XM_006235451.2 | XP_006235513.1 | 6, 7 | 670 |
Gene Symbol: Nfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053462.2 | NP_445914.2 | 671 | |||
XM_006235365.2 | XP_006235427.1 | 670 | ||||
GenBank mRNA | AF336041.1 | - | 632 | |||
BC089205.1 | - | 637 | ||||
BC104699.1 | - | 671 | 7189705 |