Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100359600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729596.4 | XP_002729642.1 | 1 | 339 |
Gene Symbol: Kpna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270802.1 | NP_001257731.1 | 3, 4 | 344 | ||
NM_053483.2 | NP_445935.2 | 3, 4 | 319 | |||
GenBank mRNA | AJ130946.1 | - | 2, 3 | 223 | ||
AY779026.1 | - | 2, 3 | 213 | |||
BC062026.1 | - | 3, 4 | 319 | 5621957 | ||
BC089787.1 | - | 3, 4 | 270 | 7326072 | ||
FM083931.1 | - | 3, 4 | 319 | |||
FM115539.1 | - | 3, 4 | 341 |