Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053499.1 | NP_445951.1 | 3 | 848 | ||
XM_006236069.2 | XP_006236131.1 | 4 | 963 | |||
XM_006236070.2 | XP_006236132.1 | 4 | 960 | |||
XM_006236071.2 | XP_006236133.1 | 3 | 835 | |||
XM_006236072.2 | XP_006236134.1 | 3 | 832 | |||
XM_006236073.2 | XP_006236135.1 | 2 | 786 | |||
GenBank mRNA | AJ292524.1 | - | 3 | 848 |