Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053520.2 | NP_445972.2 | 7 | 1082 | ||
XM_006252347.2 | XP_006252409.1 | 8 | 2365 | |||
XM_006252348.2 | XP_006252410.1 | 7 | 1103 | |||
XM_006252349.2 | XP_006252411.1 | 9 | 2531 | |||
XM_006252351.2 | XP_006252413.1 | 8 | 1609 | |||
XM_006252352.2 | XP_006252414.1 | 8 | 1569 | |||
XM_006252354.2 | XP_006252416.1 | 7 | 2257 | |||
XM_008770898.1 | XP_008769120.1 | 8 | 3869 | |||
XM_008770899.1 | XP_008769121.1 | 8 | 1565 | |||
XM_008770900.1 | XP_008769122.1 | 8 | 1121 | |||
XM_008770901.1 | XP_008769123.1 | 8 | 3845 | |||
XM_008770902.1 | XP_008769124.1 | 7 | 3761 | |||
XM_008770903.1 | XP_008769125.1 | 7 | 3737 | |||
GenBank mRNA | AB030216.1 | - | 6 | 743 | ||
AB030217.1 | - | 5 | 578 | |||
AB046531.1 | - | 7 | 861 | |||
FQ221456.1 | - | 9 | 1650 |