Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053570.1 | NP_446022.1 | 7 | 955 | ||
XM_006248000.2 | XP_006248062.1 | 7 | 1056 | |||
GenBank mRNA | AF109643.1 | - | 7 | 884 | ||
AF109644.1 | - | 7 | 884 | |||
BC088313.1 | - | 7 | 955 | 7324723 |