Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec31a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033021.1 | NP_148981.1 | 15 | 1943 | ||
XM_006250676.2 | XP_006250738.1 | 15 | 1988 | |||
XM_006250677.1 | XP_006250739.1 | 15 | 1721 | |||
XM_006250678.2 | XP_006250740.1 | 15 | 1988 | |||
XM_006250679.2 | XP_006250741.1 | 14 | 1910 | |||
XM_006250680.2 | XP_006250742.1 | 15 | 1988 | |||
XM_006250681.2 | XP_006250743.1 | 15 | 1988 | |||
XM_006250682.2 | XP_006250744.1 | 13 | 1871 | |||
XM_006250683.2 | XP_006250745.1 | 15 | 1988 | |||
XM_006250684.2 | XP_006250746.1 | 15 | 1988 | |||
XM_006250685.2 | XP_006250747.1 | 13 | 1871 | |||
XM_006250686.2 | XP_006250748.1 | 13 | 1871 | |||
XM_006250687.2 | XP_006250749.1 | 15 | 1988 | |||
XM_006250688.2 | XP_006250750.1 | 15 | 1988 | |||
XM_006250689.2 | XP_006250751.1 | 13 | 1871 | |||
XM_006250690.1 | XP_006250752.1 | 13 | 1605 | |||
GenBank mRNA | AF034582.1 | - | 15 | 1943 | ||
BC085722.1 | - | 13 | 1611 | 7107486 |