Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gosr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053584.2 | NP_446036.1 | 4, 5 | 340 | ||
XM_006246914.1 | XP_006246976.1 | 4, 5 | 408 | |||
XM_008768132.1 | XP_008766354.1 | 3, 4 | 211 | |||
GenBank mRNA | BC126068.1 | - | 4, 5 | 344 | 7387185 |