Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033968.1 | NP_001029140.1 | 9 | 1049 | ||
NM_053609.2 | NP_446061.2 | 9 | 1207 | |||
XM_006256082.1 | XP_006256144.1 | 9 | 1111 | |||
XM_006256083.1 | XP_006256145.1 | 9 | 1111 | |||
XM_006256084.1 | XP_006256146.1 | 8 | 1057 | |||
XM_006256085.1 | XP_006256147.1 | 9 | 1111 | |||
XM_006256086.2 | XP_006256148.1 | 9 | 1247 | |||
XM_006256087.2 | XP_006256149.1 | 9 | 1324 | |||
XM_006256088.1 | XP_006256150.1 | 9 | 1111 | |||
XM_006256089.1 | XP_006256151.1 | 9 | 1111 | |||
XM_006256090.1 | XP_006256152.1 | 8 | 1057 | |||
XM_006256091.1 | XP_006256153.1 | 8 | 1057 | |||
XM_006256092.1 | XP_006256154.1 | 9 | 1111 | |||
XM_006256093.2 | XP_006256155.1 | 8 | 1195 | |||
XM_006256094.1 | XP_006256156.1 | 9 | 1111 | |||
GenBank mRNA | AB018791.1 | - | 9 | 1163 | ||
BC100141.1 | - | 9 | 1049 | 7315623 | ||
FQ223705.1 | - | 8 | 1170 |