Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rraga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053973.2 | NP_446425.1 | 1 | 634 | ||
GenBank mRNA | BC061850.1 | - | 1 | 635 | 5621998 | |
X85183.1 | - | 1 | 619 |