Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacsin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130740.2 | NP_570096.2 | 9 | 1349 | ||
XM_006242049.2 | XP_006242111.1 | 8 | 1567 | |||
XM_006242050.2 | XP_006242112.1 | 10 | 2048 | |||
XM_006242051.1 | XP_006242113.1 | 10 | 1297 | |||
XM_006242052.2 | XP_006242114.1 | 10 | 1958 | |||
XM_006242053.1 | XP_006242115.1 | 10 | 1320 | |||
XM_006242055.1 | XP_006242117.1 | 9 | 1291 | |||
XM_008765660.1 | XP_008763882.1 | 9 | 5796 | |||
XM_008765661.1 | XP_008763883.1 | 9 | 1915 | |||
XM_008765662.1 | XP_008763884.1 | 9 | 1155 | |||
GenBank mRNA | AF139492.1 | - | 10 | 1273 | ||
AF139494.1 | - | 9 | 1267 | |||
BC070911.1 | - | 9 | 1349 | 7104542 | ||
FQ230278.1 | - | 2 | 117 |