Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134457.1 | NP_604452.1 | 2 | 901 | ||
GenBank mRNA | AB067815.1 | - | 2 | 901 | ||
AF389477.1 | - | 2 | 455 |