Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134371.2 | NP_599198.2 | 18 | 2414 | ||
XM_008767192.1 | XP_008765414.1 | 19 | 2676 | |||
XM_008767193.1 | XP_008765415.1 | 19 | 2676 | |||
GenBank mRNA | AY072788.2 | - | 18 | 2414 |