Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270630.1 | NP_001257559.1 | 2 | 1030 | ||
NM_001270631.1 | NP_001257560.1 | 2 | 1181 | |||
NM_001270632.1 | NP_001257561.1 | 2 | 1098 | |||
NM_001270633.1 | NP_001257562.1 | 2 | 1022 | |||
NM_001270634.1 | NP_001257563.1 | 3 | 909 | |||
NM_001270635.1 | NP_001257564.1 | 2 | 908 | |||
NM_001270636.1 | NP_001257565.1 | 2 | 894 | |||
NM_001270637.1 | NP_001257566.1 | 2 | 886 | |||
NM_001270638.1 | NP_001257567.1 | 2 | 763 | |||
NM_012513.4 | NP_036645.2 | 2 | 1321 | |||
XM_006234684.2 | XP_006234746.1 | 2 | 1029 | |||
XM_008762078.1 | XP_008760300.1 | 2 | 2955 | |||
GenBank mRNA | AY559248.1 | - | 2 | 824 | ||
AY559249.1 | - | 2 | 838 | |||
AY559250.1 | - | 3 | 910 | |||
BC087634.1 | - | 2 | 1016 | 7319966 | ||
EF125675.1 | - | 2 | 1321 | |||
EF125676.1 | - | 2 | 887 | |||
EF125677.1 | - | 2 | 1099 | |||
EF125678.1 | - | 2 | 1182 | |||
EF125679.1 | - | 2 | 1022 | |||
EF125680.1 | - | 2 | 1030 | |||
EF125686.1 | - | 2 | 908 | |||
EF125687.1 | - | 2 | 763 | |||
EF125688.1 | - | 2 | 895 | |||
EF125689.1 | - | 2 | 959 | |||
EF125690.1 | - | 1 | 1153 | |||
KC855561.1 | - | 1 | 191 | |||
M61175.1 | - | 2 | 733 | |||
X67108.1 | - | 1 | 682 |