Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012548.2 | NP_036680.1 | 5 | 729 | ||
GenBank mRNA | AF122903.1 | - | 3 | 275 | ||
M64711.1 | - | 5 | 724 |