Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769997.1 | XP_008768219.1 | 2 | 1114 | ||
XM_008769999.1 | XP_008768221.1 | 2 | 768 | |||
XM_008770000.1 | XP_008768222.1 | 2 | 768 |
Gene Symbol: Ugt2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031533.5 | NP_113721.4 | 2 | 774 | ||
GenBank mRNA | BC089792.1 | - | 2 | 757 | 7370057 | |
DQ351215.1 | - | 2 | 1115 | |||
EX489691.1 | - | 2 | 341 | |||
X03478.1 | - | 2 | 663 |