Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031543.1 | NP_113731.1 | 8 | 1201 | ||
XM_006230520.1 | XP_006230582.1 | 9 | 1384 | |||
GenBank mRNA | AF056333.1 | - | 3 | 283 | ||
AF061442.1 | - | 8 | 1189 | |||
BC081774.1 | - | 8 | 1203 | 7110096 | ||
FQ209397.1 | - | 8 | 1220 | |||
FQ209597.1 | - | 3 | 320 | |||
FQ210615.1 | - | 8 | 1130 | |||
FQ210662.1 | - | 3 | 320 | |||
FQ210815.1 | - | 8 | 1222 | |||
FQ211086.1 | - | 8 | 1222 | |||
FQ212595.1 | - | 5 | 567 | |||
FQ218528.1 | - | 6 | 840 | |||
FQ218532.1 | - | 8 | 1201 | |||
FQ219179.1 | - | 5 | 567 | |||
FQ219736.1 | - | 8 | 1220 | |||
J02627.1 | - | 8 | 1201 |