Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031543.1 | NP_113731.1 | 9 | 1432 | ||
XM_006230520.1 | XP_006230582.1 | 10 | 1615 | |||
GenBank mRNA | AF061442.1 | - | 9 | 1420 | ||
BC081774.1 | - | 9 | 1434 | 7110096 | ||
FQ209397.1 | - | 9 | 1451 | |||
FQ209597.1 | - | 4 | 551 | |||
FQ210662.1 | - | 4 | 551 | |||
FQ210815.1 | - | 9 | 1453 | |||
FQ211086.1 | - | 9 | 1453 | |||
FQ212595.1 | - | 6 | 798 | |||
FQ218528.1 | - | 7 | 1071 | |||
FQ218532.1 | - | 9 | 1432 | |||
FQ219179.1 | - | 6 | 798 | |||
FQ219736.1 | - | 9 | 1451 | |||
J02627.1 | - | 9 | 1432 |