Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012804.1 | NP_036936.1 | 5, 6 | 439 | ||
GenBank mRNA | D90038.1 | - | 5, 6 | 439 |