Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142366.2 | NP_001135838.1 | 5 | 1019 | ||
NM_001270558.2 | NP_001257487.1 | 4 | 846 | |||
NM_001270559.2 | NP_001257488.1 | 4 | 854 | |||
NM_012825.4 | NP_036957.1 | 5 | 1011 | |||
GenBank mRNA | L27588.1 | - | 4 | 879 | ||
U14007.1 | - | 5 | 945 |