Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033715.1 | NP_001028887.1 | 8, 9 | 695 | ||
NM_001033716.1 | NP_001028888.1 | 7, 8 | 626 | |||
NM_053295.2 | NP_445747.2 | 9, 10 | 809 | |||
XM_006231698.2 | XP_006231760.1 | 9, 10 | 1217 | |||
XM_006231699.2 | XP_006231761.1 | 8, 9 | 942 | |||
GenBank mRNA | AF346597.1 | - | 5, 6 | 307 | ||
AF346598.1 | - | 7, 8 | 490 | |||
BC091239.1 | - | 8, 9 | 695 | 7324026 | ||
DQ186624.1 | - | 9, 10 | 1003 | |||
DQ186625.1 | - | 7, 8 | 820 | |||
DQ186626.1 | - | 8, 9 | 934 | |||
DQ186627.1 | - | 8, 9 | 889 | |||
DQ186628.1 | - | 7, 8 | 850 | |||
DQ186629.1 | - | 6, 7 | 781 | |||
DQ287975.1 | - | 2, 3 | 681 | |||
Y13587.1 | - | 6, 7 | 388 | |||
Y13588.1 | - | 7, 8 | 502 | |||
Y13589.1 | - | 5, 6 | 319 | |||
Y13591.1 | - | 6, 7 | 388 |