Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033236.1 | NP_150239.1 | 8 | 742 | ||
GenBank mRNA | D50694.1 | - | 8 | 742 |