Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013166.1 | NP_037298.1 | 1 | 177 | ||
GenBank mRNA | FQ216326.1 | - | 1 | 140 | ||
X17457.1 | - | 1 | 177 |