Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145880.3 | NP_665887.3 | 5 | 1736 | ||
GenBank mRNA | FQ213041.1 | - | 4 | 1007 | ||
S71523.1 | - | 5 | 1736 |