Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153621.1 | NP_705885.1 | 12 | 1517 | ||
XM_008763854.1 | XP_008762076.1 | 15 | 2177 | |||
GenBank mRNA | AB072426.1 | - | 12 | 1517 |