Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153621.1 | NP_705885.1 | 13 | 1639 | ||
XM_008763854.1 | XP_008762076.1 | 16 | 2299 | |||
GenBank mRNA | AB072426.1 | - | 13 | 1639 |