Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105768.1 | NP_001099238.1 | 5 | 663 | ||
GenBank mRNA | BC168712.1 | - | 5 | 666 | 8366833 |