Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phospho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105833.1 | NP_001099303.1 | 3 | 1180 | ||
XM_006247184.2 | XP_006247246.1 | 3 | 1043 | |||
XM_008767970.1 | XP_008766192.1 | 3 | 1613 |