Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033683.1 | NP_001028855.1 | 2 | 464 | ||
GenBank mRNA | BC101882.1 | - | 2 | 464 | 7929680 |