Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sppl2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105847.1 | NP_001099317.1 | 1 | 1752 | ||
XM_006247504.2 | XP_006247566.1 | 1 | 1748 | |||
GenBank mRNA | BC160911.1 | - | 1 | 1721 | 7456885 |