Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105913.1 | NP_001099383.1 | 2 | 1185 | ||
XM_006248924.2 | XP_006248986.1 | 2 | 953 | |||
XM_006248926.2 | XP_006248988.1 | 2 | 913 | |||
XM_008768949.1 | XP_008767171.1 | 2 | 2334 |