Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105981.1 | NP_001099451.1 | 5 | 504 | ||
XM_008769819.1 | XP_008768041.1 | 4 | 352 |