Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013048.1 | NP_001013066.1 | 3, 4 | 721 | ||
GenBank mRNA | BC086582.1 | - | 3, 4 | 721 | 7307584 |